Assay Details
Target Gene Details
Entrez Gene ID: | 268451 |
Gene Name: | RAB11 family interacting protein 4 (class II) |
Gene Aliases: |
A730072L08Rik, RAB11-FIP4, mKIAA1821 |
Location: |
Chr.11:79591021-79698023 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip4 | NM_175543.3 | 15 | 2505 | NP_780752.1 |
| AK046734.1 | 15 | 2507 | BAC32850.1 | |
| AK122559.1 | 7 | 1341 | BAC65841.1 | |
| AK147219.1 | 15 | 2577 | ||
| AK147344.1 | 15 | 2701 |

Genomic Map