Assay Details
Target Gene Details
Entrez Gene ID: | 19324 |
Gene Name: | RAB1, member RAS oncogene family |
Gene Aliases: |
Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012 |
Location: |
Chr.11:20201602-20226856 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab1 | NM_008996.3 | 6 | 2294 | NP_033022.1 |
| AF226873.1 | 6 | 2294 | AAF33844.1 | |
| AK078671.1 | 1 | 1257 | ||
| AK129477.1 | 6 | 2452 | BAC98287.1 | |
| AK152978.1 | 6 | 2463 | ||
| AK159425.1 | 6 | 2458 | BAE35072.1 | |
| AK166249.1 | 6 | 2455 | BAE38660.1 |

Genomic Map