Assay Details
Target Gene Details
Entrez Gene ID: | 19376 |
Gene Name: | RAB34, member RAS oncogene family |
Gene Aliases: |
AI326479, Narr, Rah, Rah1 |
Location: |
Chr.11:78187879-78192193 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab34 | NM_001159482.1 | 11 | 1300 | NP_001152954.1 |
| NM_033475.3 | 10 | 1645 | NP_258436.2 | |
| AB082927.1 | 10 | 1558 | BAC16803.1 | |
| AF327929.1 | 11 | 1250 | AAK11241.1 | |
| AK197089.1 | 1 | 244 | ||
| AK207859.1 | 1 | 244 | ||
| AK211856.1 | 1 | 244 | ||
| AK211857.1 | 1 | 244 | ||
| AK217557.1 | 1 | 244 | ||
| BC038638.1 | 10 | 1120 | AAH38638.1 | |
| S72304.1 | 8 | 903 | AAB20669.1 |

Genomic Map