Assay Details
Target Gene Details
Entrez Gene ID: | 20370 |
Gene Name: | seizure related gene 6 |
Gene Aliases: |
BSRP-C, D11Bhm177e, sez-6 |
Location: |
Chr.11:77930839-77979052 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sez6 | NM_001291225.1 | 16 | 3280 | NP_001278154.1 |
| NM_021286.4 | 17 | 3335 | NP_067261.2 | |
| AK147705.1 | 13 | 3825 | ||
| AK161576.1 | 12 | 1745 | BAE36472.1 | |
| AK165757.1 | 15 | 3394 | ||
| AK220513.1 | 16 | 3965 | BAD90303.1 | |
| BC053011.1 | 17 | 3231 | AAH53011.1 | |
| BC055345.1 | 15 | 3658 | AAH55345.1 | |
| D29763.1 | 16 | 3107 | BAA06167.1 |

Genomic Map