Assay Details
Target Gene Details
Entrez Gene ID: | 67465 |
Gene Name: | splicing factor 3a, subunit 1 |
Gene Aliases: |
1200014H24Rik, 5930416L09Rik, AI159724 |
Location: |
Chr.11:4160354-4182541 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3a1 | NM_026175.5 | 16 | 3661 | NP_080451.4 |
| AK020032.1 | 1 | 132 | ||
| AK030115.1 | 8 | 2187 | ||
| BC010727.1 | 7 | 2116 | AAH10727.1 |
More Information
Set Membership: |
|

Genomic Map