Assay Details
Target Gene Details
Entrez Gene ID: | 20742 |
Gene Name: | spectrin beta, non-erythrocytic 1 |
Gene Aliases: |
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049 |
Location: |
Chr.11:30099395-30268204 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptbn1 | NM_009260.2 | 22 | 5165 | NP_033286.2 |
| NM_175836.2 | 22 | 4825 | NP_787030.2 | |
| AF016040.1 | AAD01566.1 | |||
| AF017112.1 | 22 | 5072 | AAD01616.1 | |
| AK220456.1 | 23 | 5027 | BAD90485.1 | |
| M74773.1 | 22 | 5145 | AAC42040.1 |

Genomic Map