Assay Details
Target Gene Details
Entrez Gene ID: | 20742 |
Gene Name: | spectrin beta, non-erythrocytic 1 |
Gene Aliases: |
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049 |
Location: |
Chr.11:30099395-30268204 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptbn1 | NM_175836.2 | 35 | 7578 | NP_787030.2 |
| AK036963.1 | 2 | 2296 | ||
| AK051630.1 | 10 | 1936 | BAC34698.1 | |
| AK079934.1 | 1 | 98 | ||
| BC024833.1 | 3 | 840 | AAH24833.1 |

Genomic Map