Assay Details
Target Gene Details
Entrez Gene ID: | 216971 |
Gene Name: | family with sequence similarity 222, member B |
Gene Aliases: |
9630020I17Rik |
Location: |
Chr.11:78094666-78157339 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam222b | NM_145430.2 | 3 | 2673 | NP_663405.1 |
| AK047860.1 | 3 | 2545 | ||
| AK085728.2 | 3 | 2680 | BAE43389.1 | |
| AK137590.1 | 1 | 1724 | BAE23423.1 | |
| AK140562.1 | 3 | 2545 | ||
| AK141119.1 | 3 | 2635 | BAE24565.1 |

Genomic Map