Assay Details
Target Gene Details
Entrez Gene ID: | 192976 |
Gene Name: | leucine rich repeat containing 75A |
Gene Aliases: |
Fam211a |
Location: |
Chr.11:62604884-62648583 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc75a | NM_198861.1 | 4 | 1527 | NP_942561.1 |
| AK146714.1 | 4 | 1216 | BAE27379.1 | |
| BC046404.1 | 4 | 1267 | AAH46404.1 | |
| BC053455.1 | 4 | 1527 | AAH53455.1 |

Genomic Map