Assay Details
Target Gene Details
Entrez Gene ID: | 15985 |
Gene Name: | CD79B antigen |
Gene Aliases: |
B29, Ig-beta, Igb, Igbeta |
Location: |
Chr.11:106311341-106314529 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd79b | NM_008339.2 | 3 | 275 | NP_032365.1 |
| AK143573.1 | 3 | 276 | BAE25444.1 | |
| BC012226.1 | 3 | 275 | AAH12226.1 | |
| CT010358.1 | 3 | 230 | CAJ18566.1 | |
| J03857.1 | 3 | 414 | AAA37274.1 |
More Information
Set Membership: |
|

Genomic Map