Assay Details
Target Gene Details
Entrez Gene ID: | 217232 |
Gene Name: | cell division cycle 27 |
Gene Aliases: |
AI452358, APC3, BC023187 |
Location: |
Chr.11:104502524-104550620 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc27 | NM_001285988.1 | 1 | 124 | NP_001272917.1 |
| NM_001285989.1 | 1 | 124 | NP_001272918.1 | |
| NM_001285990.1 | 1 | 124 | NP_001272919.1 | |
| NM_145436.2 | 1 | 124 | NP_663411.2 | |
| AK148278.1 | ||||
| AK161471.1 | ||||
| BC023187.1 | AAH23187.1 | |||
| BC157955.1 | 1 | 123 | ||
| BC172100.1 | 1 | 123 | ||
| BY013739.1 | ||||
| BY036319.1 | 1 | 125 |
Target Gene Details
Entrez Gene ID: | 17896 |
Gene Name: | myosin, light polypeptide 4 |
Gene Aliases: |
ALC1, AMLC, ELC, ELC1a, GT1, MLC1a, Myla |
Location: |
Chr.11:104550530-104587219 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl4 |
More Information
Set Membership: |
|

Genomic Map