Assay Details
Target Gene Details
Entrez Gene ID: | 14406 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2 |
Gene Aliases: |
GABAA-R, Gabrg-2, gamma2 |
Location: |
Chr.11:41910190-42000840 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabrg2 | NM_008073.3 | 1 | 419 | NP_032099.1 |
| NM_177408.6 | 1 | 419 | NP_803127.3 | |
| AF233775.1 | AAK71573.1 | |||
| AF233776.1 | AAK71574.1 | |||
| AF233777.1 | AAK71575.1 | |||
| AF233778.1 | AAK71576.1 | |||
| AF233779.1 | AAK71577.1 | |||
| AF233780.1 | AAK71578.1 | |||
| AF233781.1 | AAK71579.1 | |||
| AF233782.1 | AAK71580.1 | |||
| AF233783.1 | AAK71581.1 | |||
| AF233784.1 | AAK71582.1 | |||
| AF233785.1 | AAK71583.1 | |||
| AF233786.1 | AAK71584.1 | |||
| AF233787.1 | AAK71585.1 | |||
| AF233788.1 | AAK71586.1 | |||
| AF233789.1 | AAK71587.1 | |||
| AF233790.1 | AAK71588.1 | |||
| AF233791.1 | AAK71589.1 | |||
| AF233792.1 | AAK71590.1 | |||
| AF233793.1 | AAK71591.1 | |||
| AF233794.1 | AAK71592.1 | |||
| AF233795.1 | AAK71593.1 | |||
| AF233796.1 | AAK71594.1 | |||
| AF233797.1 | AAK71595.1 | |||
| AF233798.1 | AAK71596.1 | |||
| AF233799.1 | AAK71597.1 | |||
| AF233800.1 | AAK71598.1 | |||
| AF233801.1 | AAK71599.1 | |||
| AF233802.1 | AAK71600.1 | |||
| AK034338.1 | 1 | 389 | ||
| AK047085.1 | BAC32956.1 | |||
| AK049857.1 | BAC33961.1 | |||
| AK136889.1 | BAE23157.1 | |||
| AK141129.1 | ||||
| BC031762.1 | 1 | 237 | AAH31762.1 | |
| M62374.1 | 1 | 383 | AAA37653.1 | |
| M86572.1 | 1 | 212 | AAB59635.1 |
More Information
Set Membership: |
|

Genomic Map