Assay Details
Target Gene Details
Entrez Gene ID: | 14406 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2 |
Gene Aliases: |
GABAA-R, Gabrg-2, gamma2 |
Location: |
Chr.11:41910190-42000840 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabrg2 | NM_008073.3 | 1 | 113 | NP_032099.1 |
| NM_177408.6 | 1 | 113 | NP_803127.3 | |
| AK034338.1 | 1 | 83 | ||
| M62374.1 | 1 | 77 | AAA37653.1 |
More Information
Set Membership: |
|

Genomic Map