Assay Details
Target Gene Details
Entrez Gene ID: | 14406 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2 |
Gene Aliases: |
GABAA-R, Gabrg-2, gamma2 |
Location: |
Chr.11:41910190-42000840 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabrg2 | NM_008073.3 | 10 | 2025 | NP_032099.1 |
| NM_177408.6 | 9 | 2001 | NP_803127.3 | |
| AK047085.1 | 10 | 1603 | BAC32956.1 | |
| AK078261.1 | 1 | 1876 | ||
| AK136889.1 | 9 | 1609 | BAE23157.1 | |
| AK141129.1 | 9 | 1644 | ||
| BC031762.1 | 10 | 1843 | AAH31762.1 | |
| M62374.1 | 10 | 1988 | AAA37653.1 | |
| M86572.1 | 9 | 1793 | AAB59635.1 |
More Information
Set Membership: |
|

Genomic Map