Assay Details
Target Gene Details
Entrez Gene ID: | 53897 |
Gene Name: | galactose-3-O-sulfotransferase 1 |
Gene Aliases: |
Cst, Gcst |
Location: |
Chr.11:3983636-3999328 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gal3st1 | NM_001177691.1 | 3 | 454 | NP_001171162.1 |
| NM_001177703.1 | 3 | 585 | NP_001171174.1 | |
| NM_016922.3 | 4 | 803 | NP_058618.2 | |
| AB032939.1 | 2 | 380 | BAA93008.1 | |
| AK002510.1 | 3 | 454 | ||
| AK007645.1 | 3 | 585 | BAB25160.1 | |
| BC026806.1 | 4 | 726 | AAH26806.1 | |
| CX239940.1 | 3 | 497 |
More Information
Set Membership: |
|

Genomic Map