Assay Details
Target Gene Details
Entrez Gene ID: | 18356 |
Gene Name: | olfactory receptor 56 |
Gene Aliases: |
IF7, MOR276-1 |
Location: |
Chr.11:49050733-49135387 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr56 | NM_010999.2 | 6 | 1199 | NP_035129.1 |
| AF102533.1 | 1 | 271 | AAD13325.1 | |
| AF102535.1 | 1 | 271 | AAD13327.1 | |
| AF102537.1 | 1 | 271 | AAD13329.1 | |
| AK053352.1 | 6 | 1199 | BAC35358.1 | |
| BC141219.1 | 5 | 1030 | ||
| BC147437.1 | 5 | 1144 |
More Information
Set Membership: |
|

Genomic Map