Assay Details
Target Gene Details
Entrez Gene ID: | 217371 |
Gene Name: | Rab40B, member RAS oncogene family |
Gene Aliases: |
SEC4L, rar |
Location: |
Chr.11:121356121-121388251 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab40b | NM_139147.3 | 6 | 1477 | NP_631886.2 |
| AK078797.1 | 1 | 331 | ||
| BC096601.1 | 6 | 1471 | AAH96601.1 |

Genomic Map