Assay Details
Target Gene Details
Entrez Gene ID: | 54189 |
Gene Name: | rabaptin, RAB GTPase binding effector protein 1 |
Gene Aliases: |
Rab5ep, rabaptin-5, rabaptin-5alpha |
Location: |
Chr.11:70844723-70943105 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabep1 | NM_001291141.1 | 17 | 4388 | NP_001278070.1 |
| NM_001291142.1 | 17 | 4397 | NP_001278071.1 | |
| NM_001291143.1 | 19 | 4588 | NP_001278072.1 | |
| NM_019400.3 | 18 | 4517 | NP_062273.2 | |
| AK163126.1 | 1 | 1756 | BAE37204.1 |

Genomic Map