Assay Details
Target Gene Details
Entrez Gene ID: | 19360 |
Gene Name: | RAD50 homolog (S. cerevisiae) |
Gene Aliases: |
Mrell, Rad50l |
Location: |
Chr.11:53649519-53707319 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad50 | NM_009012.2 | 25 | 4965 | NP_033038.2 |
| AK132949.1 | 5 | 1805 | BAE21438.1 | |
| U66887.1 | 25 | 4943 | AAC52894.1 |

Genomic Map