Assay Details
Target Gene Details
Entrez Gene ID: | 170720 |
Gene Name: | caspase recruitment domain family, member 14 |
Gene Aliases: |
Bimp2, CARMA2 |
Location: |
Chr.11:119307096-119345375 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card14 | NM_130886.3 | 22 | 3745 | NP_570956.1 |
| AF363457.1 | AAK60137.1 | |||
| AK028888.1 | 16 | 3130 | BAC26175.1 | |
| BC004692.1 | AAH04692.1 | |||
| BC029102.1 | 22 | 3745 | AAH29102.1 |
Target Gene Details
Entrez Gene ID: | 27029 |
Gene Name: | N-sulfoglucosamine sulfohydrolase (sulfamidase) |
Gene Aliases: |
4632406A19Rik |
Location: |
Chr.11:119343489-119355510 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgsh | NM_018822.3 | 8 | 2651 | NP_061292.2 |
| AK034220.1 | 8 | 2455 | ||
| AK052266.1 | 8 | 2644 | BAC34906.1 | |
| AK076267.1 | 8 | 2681 | ||
| AK128926.1 | 8 | 2559 | BAC87657.1 |
More Information
Set Membership: |
|

Genomic Map