Assay Details
Target Gene Details
Entrez Gene ID: | 20445 |
Gene Name: | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 |
Gene Aliases: |
Siat7a |
Location: |
Chr.11:116765025-116775507 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6galnac1 | NM_011371.2 | 9 | 2096 | NP_035501.2 |
| Y11274.1 | 9 | 2096 | CAA72137.1 |

Genomic Map