Assay Details
Target Gene Details
Entrez Gene ID: | 218333 |
Gene Name: | interactor of little elongation complex ELL subunit 1 |
Gene Aliases: |
C77245, mKIAA0947 |
Location: |
Chr.13:70588688-70637838 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ice1 | NM_144837.3 | 13 | 4254 | NP_659086.2 |
| AK129244.1 | 1 | 2691 | BAC98054.1 | |
| AK134564.1 | 1 | 697 |
More Information
Set Membership: |
|

Genomic Map