Assay Details
Target Gene Details
Entrez Gene ID: | 109181 |
Gene Name: | thyroid hormone receptor interactor 11 |
Gene Aliases: |
2610511G22Rik, 3110031G15Rik, 6030460N08Rik, AI450776, GMAP-210, TRIP230 |
Location: |
Chr.12:101834043-101913649 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip11 | NM_028446.1 | 7 | 1356 | NP_082722.1 |
| AK014103.1 | BAB29158.1 | |||
| AK077954.1 | 7 | 1306 | BAC37082.1 | |
| AK081763.1 | 7 | 1269 | ||
| AK148294.1 | 7 | 1332 | BAE28462.1 | |
| BC158098.1 | 7 | 1452 |
More Information
Set Membership: |
|

Genomic Map