Assay Details
Target Gene Details
Entrez Gene ID: | 57441 |
Gene Name: | geminin |
Gene Aliases: |
AW546347, Gem |
Location: |
Chr.13:24751845-24761937 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gmnn | NM_020567.2 | NP_065592.1 | ||
| AF068780.1 | AAC24228.1 | |||
| AK131677.1 | 1 | 119 | ||
| AK131959.1 | BAE20903.1 | |||
| BC020061.1 | 1 | 179 | AAH20061.1 | |
| BY045588.1 |
More Information
Set Membership: |
|

Genomic Map