Assay Details
Target Gene Details
Entrez Gene ID: | 77134 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A0 |
Gene Aliases: |
1110055B05Rik, 3010025E17Rik, Hnrpa0, hnRNP A0 |
Location: |
Chr.13:58125879-58128556 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpa0 | NM_029872.1 | 1 | 1788 | NP_084148.1 |
| AK004260.1 | 1 | 94 | ||
| AK172180.1 | 1 | 302 |

Genomic Map