Assay Details
Target Gene Details
Entrez Gene ID: | 70078 |
Gene Name: | nucleolar protein 7 |
Gene Aliases: |
2210008F15Rik, 5730556I21Rik, NOP27, RARG-1 |
Location: |
Chr.13:43398376-43402858 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol7 | NM_023554.2 | 1 | 52 | NP_076043.2 |
| AB041566.1 | BAA95050.1 | |||
| AK008670.1 | BAB25823.1 | |||
| AK019972.1 | BAB31943.1 | |||
| AK032963.1 | BAC28101.1 | |||
| BC090637.1 | ||||
| BC130012.1 | ||||
| BY099842.1 | 1 | 52 |

Genomic Map