Assay Details
Target Gene Details
Entrez Gene ID: | 28126 |
Gene Name: | NOP16 nucleolar protein |
Gene Aliases: |
AA409471, D13Wsu177e |
Location: |
Chr.13:54584191-54590074 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nop16 | NM_178605.4 | 5 | 1328 | NP_848720.1 |
| AK013251.1 | 5 | 1329 | BAB28746.1 |

Genomic Map