Assay Details
Target Gene Details
Entrez Gene ID: | 277154 |
Gene Name: | NYN domain and retroviral integrase containing |
Gene Aliases: |
AA409373, C80530, mKIAA1305 |
Location: |
Chr.14:55854115-55874736 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nynrin | NM_001040072.1 | 2 | 857 | NP_001035161.1 |
| BC151043.1 | 3 | 879 |
More Information
Set Membership: |
|

Genomic Map