Assay Details
Target Gene Details
Entrez Gene ID: | 108671 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 9 |
Gene Aliases: |
5330419I01Rik, AU020082 |
Location: |
Chr.14:20384638-20388910 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc9 | NM_134081.5 | 3 | 659 | NP_598842.1 |
| AK043725.1 | 3 | 629 | BAC31630.1 | |
| AK075979.1 | 3 | 620 | BAC36092.1 | |
| AK077324.1 | 3 | 629 | BAC36750.1 | |
| AK153033.1 | 3 | 619 | BAE31665.1 | |
| AK162075.1 | 3 | 534 | BAE36712.1 | |
| BC014686.1 | 3 | 621 | AAH14686.2 | |
| BC023787.1 | 3 | 617 | AAH23787.1 | |
| BC027012.1 | 3 | 612 | AAH27012.1 | |
| CX238858.1 | 3 | 659 |
More Information
Set Membership: |
|

Genomic Map