Assay Details
Target Gene Details
Entrez Gene ID: | 108671 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 9 |
Gene Aliases: |
5330419I01Rik, AU020082 |
Location: |
Chr.14:20384638-20388910 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc9 | NM_134081.5 | 5 | 1504 | NP_598842.1 |
| AK077324.1 | 5 | 1474 | BAC36750.1 |
More Information
Set Membership: |
|

Genomic Map