Assay Details
Target Gene Details
Entrez Gene ID: | 212483 |
Gene Name: | family with sequence similarity 193, member B |
Gene Aliases: |
IRIZIO, Kiaa1931 |
Location: |
Chr.13:55539316-55571120 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam193b | NM_145382.4 | 8 | 3101 | NP_663357.2 |
| AK040568.1 | 2 | 1229 | BAC30629.1 | |
| AK155237.1 | 8 | 3095 | BAE33140.1 | |
| AK173303.1 | 8 | 1950 | BAD32581.1 | |
| BC021381.1 | 2 | 691 | AAH21381.1 | |
| BC023015.1 | 2 | 633 | AAH23015.1 | |
| BC025483.1 | 8 | 1960 | AAH25483.2 |

Genomic Map