Assay Details
Target Gene Details
Entrez Gene ID: | 13371 |
Gene Name: | deiodinase, iodothyronine, type II |
Gene Aliases: |
5DII, AI324267, DIOII |
Location: |
Chr.12:90724552-90738438 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dio2 | NM_010050.2 | 1 | 250 | NP_034180.1 |
| AF093137.1 | 1 | 108 | AAF00069.2 | |
| AF096875.1 | 1 | 250 | AAD11422.1 | |
| AF177196.1 | 1 | 120 | AAD53113.1 | |
| AF177197.1 | 1 | 120 | AAD53114.1 | |
| AK045607.1 | ||||
| BC125383.1 | 1 | 145 | ||
| BC125385.1 | 1 | 145 |

Genomic Map