Assay Details
Target Gene Details
Entrez Gene ID: | 194655 |
Gene Name: | Kruppel-like factor 11 |
Gene Aliases: |
9830142A17, D12Ertd427e, Tieg2, Tieg2b, Tieg3 |
Location: |
Chr.12:24651371-24662782 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf11 | NM_178357.3 | 2 | 238 | NP_848134.1 |
| AJ459773.2 | 2 | 84 | CAD30835.1 | |
| AK036618.1 | 2 | 224 | BAC29509.1 | |
| AK038397.1 | 2 | 336 | BAC29984.1 | |
| AK050156.1 | 2 | 239 | BAC34099.1 | |
| AK054170.1 | 2 | 210 | BAC35678.1 | |
| AK082838.1 | 2 | 227 | BAC38647.1 | |
| AK084913.1 | 2 | 211 | BAC39307.1 | |
| AK085319.1 | 2 | 210 | ||
| AK089044.1 | 2 | 218 | BAC40717.1 | |
| AK161233.1 | 2 | 230 | BAE36256.1 | |
| BC060642.1 | 2 | 236 | AAH60642.1 |
More Information
Set Membership: |
|

Genomic Map