Assay Details
Target Gene Details
Entrez Gene ID: | 26556 |
Gene Name: | homer homolog 1 (Drosophila) |
Gene Aliases: |
PSD-Zip45, SYN47, Ves-1, homer-1, vesl-1 |
Location: |
Chr.13:93304495-93405129 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Homer1 | NM_001284189.1 | 9 | 2652 | NP_001271118.1 |
| NM_147176.3 | 10 | 2688 | NP_671705.2 | |
| NM_152134.3 | 9 | 2171 | NP_687036.1 | |
| AK076543.1 | 4 | 1221 | BAC36386.1 | |
| AY137385.1 | 9 | 2171 | AAM95461.1 |

Genomic Map