Assay Details
Target Gene Details
Entrez Gene ID: | 75607 |
Gene Name: | WNK lysine deficient protein kinase 2 |
Gene Aliases: |
1810073P09Rik, AW122246, ESTM15, X83337, mKIAA1760 |
Location: |
Chr.13:49036302-49148574 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wnk2 | NM_001290311.1 | 8 | 2351 | NP_001277240.1 |
| NM_001290313.1 | 8 | 2351 | NP_001277242.1 | |
| NM_029361.4 | 8 | 2351 | NP_083637.2 | |
| AK147550.1 | 8 | 2038 | BAE27991.1 | |
| BC046464.1 | 2 | 480 | AAH46464.1 |
More Information
Set Membership: |
|

Genomic Map