Assay Details
Target Gene Details
Entrez Gene ID: | 105638 |
Gene Name: | diphthamine biosynthesis 3 |
Gene Aliases: |
DELGIP1, DelgipP1, Desr1, KTI11, Zcsl2 |
Location: |
Chr.14:32080517-32085692 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dph3 | NM_001047433.2 | 1 | 74 | NP_001040898.1 |
| NM_001284346.1 | 1 | 74 | NP_001271275.1 | |
| NM_172254.4 | 1 | 74 | NP_758458.1 | |
| NR_104303.1 | 1 | 74 | ||
| NR_104304.1 | 1 | 74 | ||
| AK011448.1 | BAC25335.1 | |||
| AK017764.1 | ||||
| AK048097.1 | BAC33240.1 | |||
| BC029910.1 | AAH29910.1 | |||
| BC039954.1 | AAH39954.1 | |||
| DN175457.1 | 1 | 48 |

Genomic Map