Assay Details
Target Gene Details
Entrez Gene ID: | 12934 |
Gene Name: | dihydropyrimidinase-like 2 |
Gene Aliases: |
AI851130, Crmp2, DRP2, Musunc33, TOAD-64, Ulip2 |
Location: |
Chr.14:66802864-66910919 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpysl2 | NM_009955.3 | 1 | 233 | NP_034085.2 |
| BC062955.1 | 1 | 233 | AAH62955.1 | |
| X87242.1 | 1 | 130 | ||
| Y10339.1 | CAA71370.1 |
More Information
Set Membership: |
|

Genomic Map