Assay Details
Target Gene Details
Entrez Gene ID: | 217378 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 27 |
Gene Aliases: |
AI639580, C330021A05Rik, Rabj, Rbj |
Location: |
Chr.12:4082568-4118013 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc27 | NM_153082.4 | 7 | 4064 | NP_694722.2 |
| AY035893.1 | AAK62027.1 | |||
| BC042643.1 | 7 | 3979 | AAH42643.1 | |
| BC051140.1 | 7 | 4026 | AAH51140.1 |

Genomic Map