Assay Details
Target Gene Details
Entrez Gene ID: | 71169 |
Gene Name: | neuroblastoma amplified sequence |
Gene Aliases: |
4933425L03Rik, Nag |
Location: |
Chr.12:13269101-13637380 on Build GRCm38 |
Assay Gene Location: | Within Intron 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nbas | NM_027706.1 | NP_081982.1 | ||
| AK146270.1 | BAE27029.1 | |||
| BC046406.1 | AAH46406.1 | |||
| BC057020.1 | AAH57020.1 | |||
| BC072598.1 | AAH72598.1 |
More Information
Set Membership: |
|

Genomic Map