Assay Details
Target Gene Details
Entrez Gene ID: | 194655 |
Gene Name: | Kruppel-like factor 11 |
Gene Aliases: |
9830142A17, D12Ertd427e, Tieg2, Tieg2b, Tieg3 |
Location: |
Chr.12:24651371-24662782 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf11 | NM_178357.3 | 4 | 2681 | NP_848134.1 |
| AK036618.1 | 4 | 2667 | BAC29509.1 | |
| AK038397.1 | 4 | 2779 | BAC29984.1 | |
| AK050156.1 | 4 | 2683 | BAC34099.1 | |
| AK054170.1 | 4 | 2653 | BAC35678.1 | |
| AK082838.1 | 4 | 2670 | BAC38647.1 | |
| AK084913.1 | 4 | 2654 | BAC39307.1 | |
| AK085319.1 | 4 | 2656 | ||
| AK089044.1 | 4 | 2661 | BAC40717.1 | |
| AK135967.1 | 1 | 776 | ||
| BC060642.1 | 4 | 2679 | AAH60642.1 |
More Information
Set Membership: |
|

Genomic Map