Assay Details
Target Gene Details
Entrez Gene ID: | 17420 |
Gene Name: | menage a trois 1 |
Gene Aliases: |
E130115E11Rik, MAT1, P36 |
Location: |
Chr.12:73112184-73273988 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnat1 |
Target Gene Details
Entrez Gene ID: | 20474 |
Gene Name: | sine oculis-related homeobox 4 |
Gene Aliases: |
AI047561, AREC3, TrexBF |
Location: |
Chr.12:73099609-73114774 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Six4 | NM_011382.2 | 1 | 756 | NP_035512.1 |
| BC137931.1 | 1 | 739 | ||
| BC137934.1 | 1 | 739 | ||
| D50416.1 | 1 | 756 | BAA08915.1 | |
| D50417.1 | 3 | 671 | BAA08916.1 | |
| D50418.1 | 3 | 529 | BAA08917.1 |
More Information
Set Membership: |
|

Genomic Map