Assay Details
Target Gene Details
Entrez Gene ID: | 15466 |
Gene Name: | histamine receptor H2 |
Gene Aliases: |
- |
Location: |
Chr.13:54191970-54236182 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrh2 | NM_001010973.2 | 3 | 2382 | NP_001010973.1 |
| AK020259.1 | 3 | 2383 | BAB32044.1 | |
| AK163760.1 | 3 | 2379 | BAE37484.1 |
More Information
Set Membership: |
|

Genomic Map