Assay Details
Target Gene Details
Entrez Gene ID: | 15466 |
Gene Name: | histamine receptor H2 |
Gene Aliases: |
- |
Location: |
Chr.13:54191970-54236182 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrh2 | NM_001010973.2 | 2 | 562 | NP_001010973.1 |
| NM_008286.2 | 2 | 562 | NP_032312.2 | |
| AB444633.1 | 1 | 59 | ||
| AB444634.1 | 1 | 59 | ||
| AF513034.1 | 1 | 59 | AAM47009.1 | |
| AF513035.1 | 1 | 59 | AAM47010.1 | |
| AK020259.1 | 2 | 563 | BAB32044.1 | |
| AK163760.1 | 2 | 559 | BAE37484.1 | |
| BB647802.1 | 2 | 559 | ||
| BC101958.1 | 1 | 57 | AAI01959.1 | |
| BC103552.1 | 1 | 58 | AAI03553.1 | |
| BC103553.1 | 1 | 58 | AAI03554.1 | |
| BC103554.1 | 1 | 58 | AAI03555.1 |
More Information
Set Membership: |
|

Genomic Map