Assay Details
Target Gene Details
Entrez Gene ID: | 320614 |
Gene Name: | RIKEN cDNA A330033J07 gene |
Gene Aliases: |
- |
Location: |
Chr.13:48044231-48262623 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A330033J07Rik | NR_102303.1 | |||
| AK034282.1 | ||||
| AK039372.1 | ||||
| AK046068.1 |
Target Gene Details
Entrez Gene ID: | 15904 |
Gene Name: | inhibitor of DNA binding 4 |
Gene Aliases: |
Idb4, bHLHb27 |
Location: |
Chr.13:48261427-48264036 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Id4 | NM_031166.2 | 1 | 335 | NP_112443.1 |
| AK041164.1 | 1 | 534 | BAC30845.1 | |
| X75018.1 | 1 | 335 | CAA52926.1 |
More Information
Set Membership: |
|

Genomic Map