Assay Details
Target Gene Details
Entrez Gene ID: | 76895 |
Gene Name: | bicaudal D homolog 2 (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.13:49341549-49387026 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bicd2 | NM_001039179.2 | 7 | 3026 | NP_001034268.1 |
| NM_001039180.2 | NP_001034269.1 | |||
| NM_029791.4 | NP_084067.1 | |||
| AJ250106.1 | CAC51393.1 | |||
| AK003456.1 | BAC25035.1 | |||
| AK122348.1 | 7 | 2999 | BAC65630.1 | |
| AK156407.1 | 7 | 2981 | ||
| AK171300.1 | BAE42379.1 | |||
| BC032198.1 | AAH32198.1 |

Genomic Map