Assay Details
Target Gene Details
Entrez Gene ID: | 11845 |
Gene Name: | ADP-ribosylation factor 6 |
Gene Aliases: |
AI788669, AW496366 |
Location: |
Chr.12:69372150-69375980 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arf6 | NM_007481.3 | 2 | 1292 | NP_031507.1 |
| AK006366.1 | 1 | 1004 | ||
| AK156960.1 | 2 | 1277 | BAE33917.1 | |
| AK179120.1 | 1 | 170 | ||
| BC003478.1 | 2 | 1254 | AAH03478.1 | |
| BC083112.1 | 2 | 1292 | AAH83112.1 | |
| D87903.1 | 2 | 1235 | BAA13495.1 |
More Information
Set Membership: |
|

Genomic Map