Assay Details
Target Gene Details
Entrez Gene ID: | 238395 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3J |
Gene Aliases: |
AI506554, EG238395, Gm4931 |
Location: |
Chr.12:104300722-104320586 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina3j | NM_001101472.2 | 4 | 1226 | NP_001094942.1 |
More Information
Set Membership: |
|

Genomic Map