Assay Details
Target Gene Details
Entrez Gene ID: | 16777 |
Gene Name: | laminin B1 |
Gene Aliases: |
C77966, C80098, C81607, D130003D08Rik, Lamb-1, Lamb1-1 |
Location: |
Chr.12:31265148-31329644 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamb1 | NM_008482.2 | 1 | 155 | NP_032508.2 |
| AK051131.1 | 1 | 156 | ||
| AK143079.1 | 1 | 157 | ||
| AK147313.1 | 1 | 154 | BAE27840.1 | |
| AK164842.1 | BAE37939.1 | |||
| BC121790.1 | 1 | 49 | ||
| BC150809.1 | 1 | 110 | ||
| CJ093872.1 | 1 | 155 | ||
| M15525.1 | 1 | 61 | AAA39407.1 |
More Information
Set Membership: |
|

Genomic Map