Assay Details
Target Gene Details
Entrez Gene ID: | 19088 |
Gene Name: | protein kinase, cAMP dependent regulatory, type II beta |
Gene Aliases: |
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta) |
Location: |
Chr.12:31958450-32061343 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkar2b | NM_011158.3 | 11 | 2674 | NP_035288.2 |
| AK081805.1 | 11 | 2451 | BAE43376.1 | |
| AK138963.1 | 11 | 2640 | BAE23838.1 | |
| AK205899.1 | 1 | 188 | ||
| AK218624.1 | 1 | 188 | ||
| BC048710.1 | AAH48710.1 | |||
| CJ238090.1 |
More Information
Set Membership: |
|

Genomic Map