Assay Details
Target Gene Details
Entrez Gene ID: | 18755 |
Gene Name: | protein kinase C, eta |
Gene Aliases: |
Pkch |
Location: |
Chr.12:73585041-73778185 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkch | NM_008856.3 | 1 | 401 | NP_032882.2 |
| AK076352.1 | 1 | 600 | ||
| AK145817.1 | 1 | 647 | BAE26670.1 | |
| AK155951.1 | 1 | 647 | BAE33517.1 | |
| AK164044.1 | 1 | 600 | BAE37603.1 | |
| BC031121.1 | 1 | 401 | AAH31121.1 | |
| D90242.1 | 1 | 338 | BAA14288.1 |
More Information
Set Membership: |
|

Genomic Map